Learn More
Enzo Life Sciences T-2 Toxin (5mg). CAS: 21259-20-1
SDP
Catalog No. 502009624
Encompass_Preferred
Mycotoxin. Induces DNA damage and apoptosis. Increases blood-brain barrier permeability and inhibits monoamine oxidase activity in the brain. Alternative name: 4beta,15-Diacetoxy-3alpha-hydroxy-8alpha-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene. Purity: ≥98% (TLC). Solubility: Soluble in methylene chloride (5mg/ml) or chloroform. Long Term Storage: -20°C.
Catalog No. 50-200-9624 Supplier Enzo Life Sciences Supplier No. ALX630101M005
Mycotoxin. Induces DNA damage and apoptosis. Increases blood-brain barrier permeability and inhibits monoamine oxidase activity in the brain. Alternative name: 4beta,15-Diacetoxy-3alpha-hydroxy-8alpha-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene. Purity: ≥98% (TLC). Solubility: Soluble in methylene chloride (5mg/ml) or chloroform. Long Term Storage: -20°C.