Learn More
Selleck Chemical LLC T-26c S0341-25MG
SDP
Catalog No. 5000041113
Encompass_Preferred
T-26c is a highly potent and selective inhibitor of matrix metalloproteinase-13 (MMP-13) with IC50 of 6 9 pM more than 2600-fold selectivity over other related metalloenzymes
Catalog No. 50-000-41113 Supplier Selleck Chemical LLC Supplier No. S034125MG
T-26c is a highly potent and selective inhibitor of matrix metalloproteinase-13 (MMP-13) with IC50 of 6 9 pM more than 2600-fold selectivity over other related metalloenzymes