Learn More
TARGETMOL CHEMICALS INC T-26C 10MG
SDP
Catalog No. 502579783
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. T-26c is highly potent and selective matrix metalloproteinase-13 (MMP-13) inhibitor (IC50 6.75 pM). purity: 100%
Catalog No. 50-257-9783 Supplier TARGETMOL CHEMICALS INC Supplier No. T536110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. T-26c is highly potent and selective matrix metalloproteinase-13 (MMP-13) inhibitor (IC50 6.75 pM). purity: 100%