Learn More
Medchemexpress LLC HY-100518 50mg Medchemexpress, T-26c CAS:869296-13-9 Purity:>98%
SDP
Catalog No. 501931864
Encompass_Preferred
Medchemexpress, HY-100518 50mg T-26c CAS:869296-13-9 T-26c is highly potent and selective matrix metalloproteinase-13 (MMP-13) inhibitor with an IC 50 of 6.75 pM and more than 2600-fold selectivity over the other related metalloenzymes [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1864 Supplier Medchemexpress LLC Supplier No. HY10051850MG
Medchemexpress, HY-100518 50mg T-26c CAS:869296-13-9 T-26c is highly potent and selective matrix metalloproteinase-13 (MMP-13) inhibitor with an IC 50 of 6.75 pM and more than 2600-fold selectivity over the other related metalloenzymes [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.