Learn More
Medchemexpress LLC HY-103085 100mg Medchemexpress, T-3775440 hydrochloride CAS:1422535-52-1 Purity:>98%
SDP
Catalog No. 501968030
Encompass_Preferred
Medchemexpress, HY-103085 100mg T-3775440 HCl CAS:1422535-52-1 T-3775440 (hydrochloride) is an irreversible lysine-specific histone demethylase (LSD1) inhibitor with an IC50 value of 2.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8030 Supplier Medchemexpress LLC Supplier No. HY103085100MG
Medchemexpress, HY-103085 100mg T-3775440 HCl CAS:1422535-52-1 T-3775440 (hydrochloride) is an irreversible lysine-specific histone demethylase (LSD1) inhibitor with an IC50 value of 2.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.