Learn More
Selleck Chemical LLC T-3775440 HCl
SDP
Supplier:  Selleck Chemical LLC S84382MG
Encompass_Preferred
T-3775440 HCl is an irreversible LSD1 inhibitor that is highly selective for LSD1 relative to other monoamine oxidases (e g MAO-A and MAO-B) with an IC50 value of 2 1 nmol/L
Catalog No. 50-000-01690