Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC T-3775440 hydrochloride | 1422535-52-1 | 99.1% | 346.85 g/mol | C18H23ClN4O | 1 ML
SDP

Catalog No. 5000246757
Encompass_Preferred

T-3775440 hydrochloride is an irreversible lysine-specific histone demethylase 1 (LSD1) inhibitor used in biochemical and cellular research to probe epigenetic regulation. The hydrochloride salt is provided as a concentrated solution for in vitro applications.

  • Irreversible LSD1 inhibitor with IC50 2.1 nM.
  • High purity (99.1%) suitable for research use.
  • Molecular weight 346.85 g/mol; formula C18H23ClN4O.
  • Supplied as 10 mM solution in DMSO, 1 mL vial.
  • Soluble in DMSO (≥ 30.18 mg/mL) and soluble in water with ultrasonic/warming.
  • Storage recommendations: in solvent, -80°C for up to 6 months or -20°C for 1 month.

Catalog No. 50-002-46757 Supplier Medchemexpress LLC Supplier No. HY10308510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T-3775440 hydrochloride is an irreversible lysine-specific histone demethylase 1 (LSD1) inhibitor used in biochemical and cellular research to probe epigenetic regulation. The hydrochloride salt is provided as a concentrated solution for in vitro applications.

  • Irreversible LSD1 inhibitor with IC50 2.1 nM.
  • High purity (99.1%) suitable for research use.
  • Molecular weight 346.85 g/mol; formula C18H23ClN4O.
  • Supplied as 10 mM solution in DMSO, 1 mL vial.
  • Soluble in DMSO (≥ 30.18 mg/mL) and soluble in water with ultrasonic/warming.
  • Storage recommendations: in solvent, -80°C for up to 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.