Learn More
Medchemexpress LLC T-3775440 hydrochloride | 1422535-52-1 | 99.1% | 346.85 g/mol | C18H23ClN4O | 10 MG
SDP
Catalog No. 5000245997
Encompass_Preferred
T-3775440 hydrochloride is the hydrochloride salt of T-3775440, an irreversible lysine-specific demethylase 1 (LSD1) inhibitor used in biochemical and cellular research. It has a reported IC50 of 2.1 nM and is supplied as a solid salt for assay and mechanistic studies.
  • Irreversible LSD1 inhibitor with reported IC50 = 2.1 nM.
  • Hydrochloride salt form suitable for biochemical and cell-based assays.
  • High purity suitable for research use (reported purity 99.13%).
  • Molecular formula C18H23ClN4O; molecular weight 346.85 g/mol.
  • Packaged in small research quantities for assay development and validation.
Catalog No. 50-002-45997 Supplier Medchemexpress LLC Supplier No. HY10308510MG
T-3775440 hydrochloride is the hydrochloride salt of T-3775440, an irreversible lysine-specific demethylase 1 (LSD1) inhibitor used in biochemical and cellular research. It has a reported IC50 of 2.1 nM and is supplied as a solid salt for assay and mechanistic studies.
  • Irreversible LSD1 inhibitor with reported IC50 = 2.1 nM.
  • Hydrochloride salt form suitable for biochemical and cell-based assays.
  • High purity suitable for research use (reported purity 99.13%).
  • Molecular formula C18H23ClN4O; molecular weight 346.85 g/mol.
  • Packaged in small research quantities for assay development and validation.