Learn More
Medchemexpress LLC T-3775440 hydrochloride | 1422535-52-1 | 99.1% | 346.85 g·mol⁻¹ | C18H23ClN4O | 25 MG
Supplier: Medchemexpress LLC HY10308525MG
T-3775440 hydrochloride is a potent, irreversible inhibitor of lysine-specific histone demethylase 1 (LSD1) with an IC50 of 2.1 nM. It shows selective inhibition of LSD1 versus MAO enzymes, reduces proliferation in multiple cell lines, and has demonstrated antitumor effects in xenograft models. Supplied as a high-purity research chemical for preclinical biochemical and cellular studies.
- Potent LSD1 inhibition (IC50 2.1 nM).
- Irreversible mechanism of action.
- High purity (~99.1%).
- Suitable for biochemical assays and cell-based studies.
- Available as solid and solution formats for flexible use.
- Stable when stored under recommended conditions.