Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC T-3775440 hydrochloride | 1422535-52-1 | 99.1% | 346.85 g/mol | C18H23ClN4O | 5 MG
SDP

Catalog No. 5000246079
Encompass_Preferred

T-3775440 hydrochloride is a small-molecule, irreversible inhibitor of lysine-specific histone demethylase 1 (KDM1/LSD1) used in biochemical and cellular research. It is supplied as a light yellow to yellow solid with high purity and characterized solubility and storage recommendations.

  • Irreversible KDM1/LSD1 inhibitor; IC50 = 2.1 nM.
  • Molecular formula C18H23ClN4O; molecular weight 346.85 g/mol.
  • Purity approximately 99.1% by HPLC.
  • Physical state: light yellow to yellow solid.
  • Solubility: DMSO ≥ 30.18 mg/mL; water 8.33 mg/mL with ultrasonic warming to 60°C.
  • Storage: powder at 4°C under inert atmosphere; solutions: -80°C (6 months), -20°C (1 month).
  • Supplied as a 5 mg quantity.

Catalog No. 50-002-46079 Supplier Medchemexpress LLC Supplier No. HY1030855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T-3775440 hydrochloride is a small-molecule, irreversible inhibitor of lysine-specific histone demethylase 1 (KDM1/LSD1) used in biochemical and cellular research. It is supplied as a light yellow to yellow solid with high purity and characterized solubility and storage recommendations.

  • Irreversible KDM1/LSD1 inhibitor; IC50 = 2.1 nM.
  • Molecular formula C18H23ClN4O; molecular weight 346.85 g/mol.
  • Purity approximately 99.1% by HPLC.
  • Physical state: light yellow to yellow solid.
  • Solubility: DMSO ≥ 30.18 mg/mL; water 8.33 mg/mL with ultrasonic warming to 60°C.
  • Storage: powder at 4°C under inert atmosphere; solutions: -80°C (6 months), -20°C (1 month).
  • Supplied as a 5 mg quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.