Learn More
Medchemexpress LLC T-3775440 hydrochloride | 1422535-52-1 | 99.1% | 346.85 g/mol | C18H23ClN4O | 50 MG
SDP
Catalog No. 5000245995
Encompass_Preferred
T-3775440 hydrochloride is an irreversible inhibitor of lysine-specific histone demethylase 1 (LSD1) with low-nanomolar potency. It is provided as a high-purity research reagent for biochemical and cellular studies of epigenetic regulation.
  • Irreversible LSD1 inhibition with an IC50 of ~2.1 nM.
  • High chemical purity suitable for research use.
  • Available in small-scale quantities for screening and mechanistic studies.
  • Useful for biochemical assays and cellular models of epigenetic regulation.
  • Provided as a hydrochloride salt to improve stability and solubility.
Catalog No. 50-002-45995 Supplier Medchemexpress LLC Supplier No. HY10308550MG
T-3775440 hydrochloride is an irreversible inhibitor of lysine-specific histone demethylase 1 (LSD1) with low-nanomolar potency. It is provided as a high-purity research reagent for biochemical and cellular studies of epigenetic regulation.
  • Irreversible LSD1 inhibition with an IC50 of ~2.1 nM.
  • High chemical purity suitable for research use.
  • Available in small-scale quantities for screening and mechanistic studies.
  • Useful for biochemical assays and cellular models of epigenetic regulation.
  • Provided as a hydrochloride salt to improve stability and solubility.