Learn More
Selleck Chemical LLC T-5224 S8966-1G
SDP
Catalog No. 5000024037
Encompass_Preferred
T-5224 is a selective inhibitor of transcription factor c-Fos/activator protein (AP)-1 with anti-inflammatory effects T-5224 specifically inhibits the DNA binding activity of c-Fos/c-Jun and the IL-1 -induced up-regulation of Mmp-3 Mmp-13 and Adamts-5 transcription
Catalog No. 50-000-24037 Supplier Selleck Chemical LLC Supplier No. S89661G
T-5224 is a selective inhibitor of transcription factor c-Fos/activator protein (AP)-1 with anti-inflammatory effects T-5224 specifically inhibits the DNA binding activity of c-Fos/c-Jun and the IL-1 -induced up-regulation of Mmp-3 Mmp-13 and Adamts-5 transcription