Learn More
Medchemexpress LLC HY-12270 50mg , T-5224 CAS:530141-72-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1227050MG
Encompass_Preferred
HY-12270 50mg T-5224 CAS: 530141-72-1 T-5224 is a transcription factor c-Fos/activator protein (AP)-1 inhibitor with anti-inflammatory effects, which specifically inhibits the DNA binding activity of c-Fos/c-Jun without affecting other transcription factors. T-5224 inhibits the IL-1β-induced up-regulation of Mmp-3, Mmp-13 and Adamts-5 transcription. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9472