Learn More
Selleck Chemical LLC T-5224 5mg
SDP
Supplier:  Selleck Chemical LLC S89665MG
Encompass_Preferred
T-5224 is a selective inhibitor of transcription factorc-Fos/activator protein (AP)-1 with anti-inflammatory effects. T-5224 specifically inhibits the DNA binding activity ofc-Fos/c-Jun and theIL-1?-induced up-regulation ofMmp-3,Mmp-13 andAdamts-5 transcription. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3459