Learn More
Medchemexpress LLC T-5342126 | 956507-49-6 | 98.1% | 457.99 g·mol-1 | C25H32ClN3O3 | 10MG
SDP
Catalog No. 5000237622
Encompass_Preferred
T-5342126 is a small-molecule toll-like receptor 4 (TLR4) antagonist for laboratory research. It inhibits LPS-induced nitric oxide production in RAW 264.7 cells (IC50 = 27.8 μM) and reduces LPS-induced IL-8, TNF-α, and IL-6 in isolated human whole blood; it also showed reduced ethanol intake and decreased microglial activation in an ethanol-dependence mouse model.
Catalog No. 50-002-37622 Supplier Medchemexpress LLC Supplier No. HY12378910MG
T-5342126 is a small-molecule toll-like receptor 4 (TLR4) antagonist for laboratory research. It inhibits LPS-induced nitric oxide production in RAW 264.7 cells (IC50 = 27.8 μM) and reduces LPS-induced IL-8, TNF-α, and IL-6 in isolated human whole blood; it also showed reduced ethanol intake and decreased microglial activation in an ethanol-dependence mouse model.