Learn More
Medchemexpress LLC T-705RMP ammonium | 2096342-42-4 | 386.23 | 1 ML
SDP
Catalog No. 5000349380
Encompass_Preferred
T-705RMP ammonium is the ammonium salt form of T-705RMP. This compound is known to exhibit weak inhibition of IMP dehydrogenase (IMPDH) activity. For optimal stability, store the product at -20°C and protect it from light. If dissolved in a solvent, it can be stored at -80°C for up to 6 months or at -20°C for up to 1 month, also protected from light.
  • Ammonium salt form of T-705RMP
  • Weakly inhibits IMP dehydrogenase (IMPDH) activity
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-49380 Supplier Medchemexpress LLC Supplier No. HY136498A10MM/1ML
T-705RMP ammonium is the ammonium salt form of T-705RMP. This compound is known to exhibit weak inhibition of IMP dehydrogenase (IMPDH) activity. For optimal stability, store the product at -20°C and protect it from light. If dissolved in a solvent, it can be stored at -80°C for up to 6 months or at -20°C for up to 1 month, also protected from light.
  • Ammonium salt form of T-705RMP
  • Weakly inhibits IMP dehydrogenase (IMPDH) activity
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light