Learn More
HIMEDIA T.A.T. Broth Base, M562, Microbiology Culture Media, Dehydrated Culture Media, Broth Base, Sterility Testing, Viscous and Cosmetic Products, Powder, 5 kg
Catalog No. NC4013734
Encompass_Preferred
T.A.T. Broth Base, prepared with added polysorbate 20, is designed for the sterility testing of highly viscous or gelatinous products such as salves, ointments and other cosmetics. Casein enzymic hydrolysate provides rich nutrition while azolectin and polysorbate 20 neutralize preservatives and antimicrobial residues that would otherwise mask contamination, final pH 7.2. The result is sensitive recovery of any surviving organisms from challenging sample matrices. Dehydrated powder, store in a cool dry place. Packaged as 5 kg.
Catalog No. NC4013734 Supplier HIMEDIA Supplier No. M5625KG
T.A.T. Broth Base, prepared with added polysorbate 20, is designed for the sterility testing of highly viscous or gelatinous products such as salves, ointments and other cosmetics. Casein enzymic hydrolysate provides rich nutrition while azolectin and polysorbate 20 neutralize preservatives and antimicrobial residues that would otherwise mask contamination, final pH 7.2. The result is sensitive recovery of any surviving organisms from challenging sample matrices. Dehydrated powder, store in a cool dry place. Packaged as 5 kg.