Learn More
World Precision Instrument T-Adapter Flow-Through Kit
SDP
Supplier:  World Precision Instrument 5399
Encompass
Do more with your ISO-NOP Nitric Oxide Sensor
This kit ncludes 2 female luer T's, 3 luer lock fittings, 3 2mm gaskets, and 6 male luer to 1/8 inch tubing fittings.
Catalog No. 50-252-1259