Learn More
Gentarget T-ANTIGEN (GFP-PURO), LENTIVIR
Catalog No. NC0845609
Encompass
Pre-made lentiviral particles expressing SV40 large T-antigen under suCMV promoter, containing GFP-puromycin fusion dual marker, provided in PBS solution.
Catalog No. NC0845609 Supplier Gentarget Supplier No. LVP016GPPBS
Pre-made lentiviral particles expressing SV40 large T-antigen under suCMV promoter, containing GFP-puromycin fusion dual marker, provided in PBS solution.