Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology T-bet/TBX21 Monoclonal Antibody, Rabbit, 100 μl
SDP

Catalog No. 502729686
Encompass

The T-bet/TBX21 (E4I2K) Rabbit Monoclonal Antibody is designed for the detection of endogenous levels of total T-bet/TBX21 protein. This recombinant monoclonal antibody is generated by immunizing animals with a recombinant protein fragment derived from mouse T-bet/TBX21. The antibody is suitable for multiple research applications, including Western blotting, immunoprecipitation, immunohistochemistry, and chromatin immunoprecipitation studies.

  • Recognizes endogenous levels of total T-bet/TBX21 protein
  • Validated for diverse applications: WB, IP, IHC (paraffin), IF (frozen), flow cytometry, ChIP, and CUT&RUN
  • Produced via immunization with a recombinant protein fragment
  • Host species: rabbit
  • Reactivity confirmed for human and mouse samples
  • Supplied in a stabilized buffer containing 50% glycerol for storage at -20°C

Catalog No. 50-272-9686 Supplier Cell Signaling Technology Supplier No. 97135S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The T-bet/TBX21 (E4I2K) Rabbit Monoclonal Antibody is designed for the detection of endogenous levels of total T-bet/TBX21 protein. This recombinant monoclonal antibody is generated by immunizing animals with a recombinant protein fragment derived from mouse T-bet/TBX21. The antibody is suitable for multiple research applications, including Western blotting, immunoprecipitation, immunohistochemistry, and chromatin immunoprecipitation studies.

  • Recognizes endogenous levels of total T-bet/TBX21 protein
  • Validated for diverse applications: WB, IP, IHC (paraffin), IF (frozen), flow cytometry, ChIP, and CUT&RUN
  • Produced via immunization with a recombinant protein fragment
  • Host species: rabbit
  • Reactivity confirmed for human and mouse samples
  • Supplied in a stabilized buffer containing 50% glycerol for storage at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.