Learn More
Medchemexpress LLC T-Boc-Aminooxy-PEG2-azide | 252378-68-0 | ≥97.0% | C11H22N4O5 | 500 MG
SDP
Catalog No. 5000353221
Encompass_Preferred
t-Boc-Aminooxy-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent due to azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups
Catalog No. 50-003-53221 Supplier Medchemexpress LLC Supplier No. HY140431500MG
t-Boc-Aminooxy-PEG2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent due to azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups