Learn More
Medchemexpress LLC t-Boc-Aminooxy-PEG4-NHS ester | 2401831-99-8 | 99.9% | C20H34N2O11 | 50 MG
SDP
Catalog No. 5000356875
Encompass_Preferred
t-Boc-Aminooxy-PEG4-NHS ester is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-56875 Supplier Medchemexpress LLC Supplier No. HY14041750MG
t-Boc-Aminooxy-PEG4-NHS ester is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only