Learn More
Medchemexpress LLC T-Boc-Met-Asp-Gly-Cys-Glu-Leu | 99.2% | 880.90 | 1 MG
SDP
Catalog No. 5000438630
Encompass_Preferred
Box5 TFA is a potent Wnt5a antagonist. It inhibits Wnt5a signaling and Wnt5a-mediated Ca2+ release, and also inhibits cell migration. It has the potential for the research of melanoma.
  • In vitro: decreases the expression of rWnt5a (0.1 μg/mL) stimulated p-MARCKS in A2058 cells.
Catalog No. 50-004-38630 Supplier Medchemexpress LLC Supplier No. HY123071A1MG
Box5 TFA is a potent Wnt5a antagonist. It inhibits Wnt5a signaling and Wnt5a-mediated Ca2+ release, and also inhibits cell migration. It has the potential for the research of melanoma.
  • In vitro: decreases the expression of rWnt5a (0.1 μg/mL) stimulated p-MARCKS in A2058 cells.