Learn More
Medchemexpress LLC T-Boc-N-amido-PEG15-Br | 3012589-40-8 | 98.0% | C37H74BrNO17 | 100 MG
SDP
Catalog No. 5000355590
Encompass_Preferred
t-Boc-N-amido-PEG15-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is intended for research use only. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55590 Supplier Medchemexpress LLC Supplier No. HY143829100MG
t-Boc-N-amido-PEG15-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is intended for research use only. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins