Learn More
Medchemexpress LLC T-Boc-N-amido-PEG15-Br | 3012589-40-8 | 98.0% | C37H74BrNO17 | 25 MG
SDP
Catalog No. 5000356972
Encompass_Preferred
t-Boc-N-amido-PEG15-Br is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56972 Supplier Medchemexpress LLC Supplier No. 25MGHY143829
t-Boc-N-amido-PEG15-Br is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins