Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich T-Boc-N-Amido-Peg2-Ch2Co2H

Catalog No. 90157750MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-157-750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-157-750MG Supplier Sigma Aldrich Supplier No. 90157750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

t-Boc-N-amido-PEG2-CH2CO2H is a heterobifunctional, PEGylated crosslinker featuring a Boc-protected amine at one end and a carboxyl group at the other. The hydrophillic PEG linker facilitates solubility in biological applications. t-Boc-N-amido-PEG2-CH2CO2H can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC™ molecules) for targeted protein degradation.PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 824426-32-6
Molecular Formula C16H22N4O10
Linear Formula C16H22N4O10
Quantity 50 mg
Synonym Ac4ManNAz
Molecular Weight (g/mol) 430.37 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.