Learn More
Medchemexpress LLC T-Boc-N-amido-PEG5-acetic acid | 890152-41-7 | 98.0% | 395.45 g/mol | C17H33NO9 | 250 MG
SDP
Catalog No. 5000280799
Encompass_Preferred
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG5 linker containing a terminal carboxylic acid and a Boc-protected amine. It is intended for research use and is commonly used as a spacer in PROTAC synthesis and bioconjugation workflows where a hydrophilic, flexible linker is required. The product is offered in multiple package sizes and is characterized by analytical data suitable for synthetic applications.
  • Boc-protected amine enabling selective deprotection.
  • Terminal carboxylic acid for straightforward amide coupling.
  • PEG5 spacer provides a hydrophilic, flexible linkage.
  • Reported analytical purity suitable for synthetic use.
  • Available in multiple package sizes for research needs.
  • Provided for research use only, not for personal use.
Catalog No. 50-002-80799 Supplier Medchemexpress LLC Supplier No. HYW190962250MG
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG5 linker containing a terminal carboxylic acid and a Boc-protected amine. It is intended for research use and is commonly used as a spacer in PROTAC synthesis and bioconjugation workflows where a hydrophilic, flexible linker is required. The product is offered in multiple package sizes and is characterized by analytical data suitable for synthetic applications.
  • Boc-protected amine enabling selective deprotection.
  • Terminal carboxylic acid for straightforward amide coupling.
  • PEG5 spacer provides a hydrophilic, flexible linkage.
  • Reported analytical purity suitable for synthetic use.
  • Available in multiple package sizes for research needs.
  • Provided for research use only, not for personal use.