Learn More
Medchemexpress LLC t-Boc-N-amido-PEG5-acetic acid | 890152-41-7 | MFCD31656900 | 98.0% | 395.45 | C17H33NO9 | 1 G
SDP
Catalog No. 5000282218
Encompass_Preferred
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG-based linker used for assembling PROTACs and other bioconjugates. It provides a flexible, hydrophilic spacer with a terminal acetic acid and a Boc-protected amine, enabling standard coupling and selective deprotection steps during synthesis.
  • Provides a five-unit polyethylene glycol spacer for flexible linkage.
  • Contains a terminal acetic acid for straightforward conjugation chemistry.
  • Features a Boc-protected amine to allow selective deprotection and functionalization.
  • Suitable for synthesis of heterobifunctional degraders and bioconjugates.
  • Available in multiple pack sizes for research-scale synthesis.
Catalog No. 50-002-82218 Supplier Medchemexpress LLC Supplier No. HYW1909621G
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG-based linker used for assembling PROTACs and other bioconjugates. It provides a flexible, hydrophilic spacer with a terminal acetic acid and a Boc-protected amine, enabling standard coupling and selective deprotection steps during synthesis.
  • Provides a five-unit polyethylene glycol spacer for flexible linkage.
  • Contains a terminal acetic acid for straightforward conjugation chemistry.
  • Features a Boc-protected amine to allow selective deprotection and functionalization.
  • Suitable for synthesis of heterobifunctional degraders and bioconjugates.
  • Available in multiple pack sizes for research-scale synthesis.