Learn More
Medchemexpress LLC T-Boc-N-amido-PEG5-tosylate | 1404111-69-8 | MFCD34596623 | 96.0% | 491.60 g/mol | C22H37NO9S | 1 G
SDP
Catalog No. 5000284131
Encompass_Preferred
A Boc-protected PEG5 tosyl linker for chemical conjugation and linker assembly. The reagent combines a hydrophilic five-unit polyethylene glycol spacer with a tosylate leaving group and a Boc-protected amine, providing improved solubility and enabling nucleophilic substitution for installation of PEG-based linkers in medicinal chemistry and bioconjugation workflows.
  • Enhances aqueous solubility via PEG5 spacer.
  • Enables nucleophilic substitution through tosylate leaving group.
  • Protects primary amine with Boc for selective deprotection.
  • Suitable for linker assembly and bioconjugation chemistry.
  • Compatible with standard organic synthesis conditions.
Catalog No. 50-002-84131 Supplier Medchemexpress LLC Supplier No. HYW4602751G
A Boc-protected PEG5 tosyl linker for chemical conjugation and linker assembly. The reagent combines a hydrophilic five-unit polyethylene glycol spacer with a tosylate leaving group and a Boc-protected amine, providing improved solubility and enabling nucleophilic substitution for installation of PEG-based linkers in medicinal chemistry and bioconjugation workflows.
  • Enhances aqueous solubility via PEG5 spacer.
  • Enables nucleophilic substitution through tosylate leaving group.
  • Protects primary amine with Boc for selective deprotection.
  • Suitable for linker assembly and bioconjugation chemistry.
  • Compatible with standard organic synthesis conditions.