Learn More
Strem, An Ascensus Company CAS# 26088-25-5. 5G. 2,4,6-Trioxa-1,3,5,7-tetramethyl-8-phosphaadamantane (~32% in xylene). MFCD27978403
SDP
Supplier:  Strem, An Ascensus Company 1513105G
Encompass
CAS# 26088-25-5. 5G. 2,4,6-Trioxa-1,3,5,7-tetramethyl-8-phosphaadamantane (~32% in xylene). MFCD27978403. Molecular Formula: C10H17O3P. Molecular Weight: 216.21. Color/Form: colorless liq. Stability: air sensitive. Strem# 15-1310. www.strem.com/catalog/v3/15-1310/
Catalog No. 50-196-3527