Learn More
Strem, An Ascensus Company CAS# 1160861-53-9. 2g. 2-(Di-t-butylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% t-BuBrettPhos. MFCD13181930
SDP
Supplier:  Strem, An Ascensus Company 1511642G
Encompass
CAS# 1160861-53-9. 2g. 2-(Di-t-butylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% t-BuBrettPhos. MFCD13181930. Molecular Weight: 484.69. Molecular Formula: C31H49O2P. Color/form: white xtl. Strem# 15-1164. http://www.strem.com/catalog/v/15-1164/
Catalog No. 50-201-6688