Learn More
Strem, An Ascensus Company CAS# 1536473-72-9. 10g. Methanesulfonato(2-(di-t-butylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), dichloromethane adduct, min. 98%[t-BuBrettPhos Palladacycle Gen. 3]. MFCD25976528
SDP
Supplier:  Strem, An Ascensus Company 46032510G
Encompass
CAS# 1536473-72-9. 10g. Methanesulfonato(2-(di-t-butylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), dichloromethane adduct, min. 98%[t-BuBrettPhos Palladacycle Gen. 3]. MFCD25976528. Molecular Weight: 854.43. Molecular Formula: C44H62NO5PPdS. Color/form: brown-green solid Strem# 46-0325. http://www.strem.com/catalog/v/46-0325/
Catalog No. 50-201-6664