Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

t-Butyl4-[4-(4,4,5,5-teramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazinecarboxylate, 90%, Thermo Scientific™

Catalog No. CC30039DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC30039DE 5 g Amber Glass Bottle
CC30039CB 250 mg Amber Glass Bottle
CC30039DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC30039DE Supplier Thermo Scientific Chemicals Supplier No. CC30039DE
Add to Cart

Chemical Identifiers

CAS 470478-90-1
Molecular Formula C21H33BN2O4
Molecular Weight (g/mol) 388.32
MDL Number MFCD06411544
InChI Key ZMAVVXGWEHZLDW-UHFFFAOYSA-N
Synonym tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate, 4-4-boc-piperazino phenylboronic acid pinacol ester, 4-4-n-boc piperazin-1-yl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid, pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-boc-piperazinyl phenylboronic acid pinacol ester, 4-n-tert-butoxycarbonylpiperazine benzeneboronic acid pinacol ester, tert-butyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate
PubChem CID 2795514
IUPAC Name tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 388.31
Chemical Name or Material tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) phenyl]piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.