Learn More
Cole-Parmer Digi-Sense Thermocouple Wire Type K 20-Gauge, High Temperature Glass Braid, 100ft
Supplier:  Cole-Parmer 0854123
Encompass_Preferred
Wire comes in 20-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
Catalog No. 50-196-1530