Learn More
Lovibond™ T-CAL™ Standard Set A, For TB 350 WL
Catalog No. 09054114 Shop All Tintometer Group Products
Change view
Click to view available options
For Use With (Equipment):
Turbidimeter
Catalog No. For Use With (Equipment)
09-054-114 Turbidimeter
1 options
This item is not returnable. View return policy
Catalog No. 09-054-114 Supplier Lovibond™ Supplier No. 194152
Add to Cart
Add to Cart
This item is not returnable. View return policy
The T-CAL™ Standard Set A, delivers all of the concentrations needed to calibrate and verify the Lovibond™ TB350 WL Portable Turbidimeter. The standards are supplied in sealed vials, eliminating the need for preparation and ensuring safe operation.
  • Supplied in ready-to-use sealed vials
  • Save time and eliminate errors - no need for dilution
  • US EPA approved as primary calibration standards
T-CAL™ Turbidity Standards from Lovibond™ are stabilized Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. All concentrations are verified to meet tight quality control specifications, are EPA compliant for reporting purposes and are guaranteed to save time.
Set includes the standards for 4 required calibration points for the Lovibond™ TB350 WL plus a 5 NTU standard for instrument verification purposes.
Specifications
Type Calibration Kit
Test Reagent Formats Sealed Glass Vials
For Use With (Equipment) Turbidimeter
Calibration Type Primary
Includes 4 Calibration Standard, 1 Verification Standard
For Use With (Application) Calibration of the Lovibond TB350 WL portable turbidimeter.
Recommended Accessories Lovibond Silicone Oil (194295) and Cleaning Cloth (197635)
Turbidity 5/20/800/2000/4000 NTU
Dimensions (L x W x H) 7.25 x 2 x 9.25 in.
Weight (English) <1 lb.
Quantity 10 mL