Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ T-CAL™ Turbidity Standard, 1.0 NTU

Catalog No. 15398362 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
1000 mL/1 L
125 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15-398-362 125 mL
15-398-363 500 mL
15-398-364 1000 mL/1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 15-398-362 Supplier Lovibond™ Supplier No. 48011112
Add to Cart
Add to Cart
This item is not returnable. View return policy

T-CAL™ Turbidity Standards are stabilized Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. Eliminate the opportunity for inaccurate turbidimeter calibrations resulting from the improper dilution or handling of Formazin stock solution.

  • Save time and eliminate errors - no need for dilution
  • EPA approved primary turbidity standard
  • Long shelf life
  • All concentrations of T-CAL™ Turbidity Standards are verified to meet tight quality control specifications, are EPA and ISO compliant for reporting purposes and guaranteed to save you time. As a primary standard, T-CAL™ Turbidity Standards can be used to calibrate or verify any manufacturer's turbidimeter.

Specifications

Type Primary Turbidity Standard
For Use With (Equipment) Turbidimeter
Calibration Type Primary
Form Liquid
For Use With (Application) Calibration or Verification of a Turbidimeter
Turbidity 1.0 NTU
Certifications/Compliance EN ISO 7027, USEPA 180.1 and APHA 2130 Part B.
Volume (Metric) 125 mL
Quantity 1 Ea.
Packaging Type Amber Plastic Bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.