Learn More
MilliporeSigma MILLIPLEX MAP Human High Sensitivity T Cell Panel Premixed 13-plex - Immunology Multiplex Assay, 96 wells
Catalog No. NC0898879
Encompass
Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Catalog No. NC0898879 Supplier MilliporeSigma Supplier No. HSTCMAG28SPMX13
Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.