Learn More
Foxx Life Sciences Polyethylene T-Type Barbed Tubing Connectors (Tee-Shaped), Three 1/8 Inch Barbs, 10/PK
Catalog No. 501178759
Encompass_Preferred
25P-2701-OEM: Connector, T Connector, Polyethylene, 1/8" Hose Barb x 1/8" Hose Barb to 1/8" Hose Barb, 10/pk. Foxx Life Sciences T connectors are available in a variety of standard sizes. Our NPT threaded and barbed tees offer strength and durability. Made from Polyethylene.
Catalog No. 50-117-8759 Supplier Foxx Life Sciences Supplier No. 25P2701OEM
25P-2701-OEM: Connector, T Connector, Polyethylene, 1/8" Hose Barb x 1/8" Hose Barb to 1/8" Hose Barb, 10/pk. Foxx Life Sciences T connectors are available in a variety of standard sizes. Our NPT threaded and barbed tees offer strength and durability. Made from Polyethylene.