Learn More
Bellco Glass, Inc. T Connector Fitting Pack, Polyethylene, 1/4" HB x 1/4" HB to 1/4" HB, 10/PK
SDP
Catalog No. 502550486
Encompass_Preferred
T connectors are available in a variety of standard sizes. Our NPT threaded and barbed tees offer strength and durability. Made from Polyethylene. Material: PolyethyleneColor: BlackLength: 1.87"OD: 0.275"ID: 0.150"
Catalog No. 50-255-0486 Supplier Bellco Glass, Inc. Supplier No. 452800276
T connectors are available in a variety of standard sizes. Our NPT threaded and barbed tees offer strength and durability. Made from Polyethylene. Material: PolyethyleneColor: BlackLength: 1.87"OD: 0.275"ID: 0.150"