Learn More
Research Products International Corp T Shaped Tubing Connectors for 3/8 Inch Tubing, Polypropylene, 12 per Bag
SDP
Catalog No. 50998417
Encompass
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power. Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Chemical-resistant polypropylene for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Connectors are autoclavable.
Catalog No. 50-998-417 Supplier Research Products International Corp Supplier No. F19606
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power. Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Chemical-resistant polypropylene for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Connectors are autoclavable.