Learn More
Sciencell Research Laboratories T/E (TRYPSIN/EDTA SOLN) 100ML
Catalog No. NC9664282
Encompass
Trypsin/EDTA solution is a sterile, phosphate and HEPES-buffered saline solution that is used to detach adherent cells from a culture surface. It contains 0.25% trypsin, 0.5 mM EDTA, 1 mM sodium pyruvate and 10 mM HEPES.
Catalog No. NC9664282 Supplier Sciencell Research Laboratories Supplier No. 0103
Trypsin/EDTA solution is a sterile, phosphate and HEPES-buffered saline solution that is used to detach adherent cells from a culture surface. It contains 0.25% trypsin, 0.5 mM EDTA, 1 mM sodium pyruvate and 10 mM HEPES.