Learn More
BIOGEMS INTERNATIONAL INC Transcription Factor Fixation/Permeabilization Concentrate
SDP
Catalog No. 501530543
Encompass
The Transcription Factor Fixation/Permeabilization solution can be used with the relevant antibodies to stain for transcription factors such as Foxp3 and intranuclear staining of cytokines, chemokines, and nuclear proteins. The concentrated solution should be used with the Transcription Factor Fixation/Permeabilization Diluent (cat#92555-00) and Permeabilization Buffer (cat# 92110-00).
Catalog No. 50-153-0543 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 925500050
The Transcription Factor Fixation/Permeabilization solution can be used with the relevant antibodies to stain for transcription factors such as Foxp3 and intranuclear staining of cytokines, chemokines, and nuclear proteins. The concentrated solution should be used with the Transcription Factor Fixation/Permeabilization Diluent (cat#92555-00) and Permeabilization Buffer (cat# 92110-00).