Learn More
Braintree Scientific T/Flex Plus Handling Gloves Extra Small
SDP
Catalog No. 501956044
Encompass
T/FLEX Plus Gloves. Cut resistant gloves for animal handling. Gloves are suitable for the safe handling of small laboratory animals including rodents. T/FLEX PLUS gloves are a 13 gauge seamless knit shell; material is Dyneema engineered fibers plated with Lycra - Spandex, Thermax and the antimicrobial treatment Alphasan combine to provide a comfortable glove featuring the highest cut resistance. Glove conform to international safety standards for cut resistance. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6044 Supplier Braintree Scientific Supplier No. TFL13G6
T/FLEX Plus Gloves. Cut resistant gloves for animal handling. Gloves are suitable for the safe handling of small laboratory animals including rodents. T/FLEX PLUS gloves are a 13 gauge seamless knit shell; material is Dyneema engineered fibers plated with Lycra - Spandex, Thermax and the antimicrobial treatment Alphasan combine to provide a comfortable glove featuring the highest cut resistance. Glove conform to international safety standards for cut resistance. **Sold for research purposes only and not for human use**