Learn More
Cayman Chemical -FluvalInate 100mg
SDP
Catalog No. 502947149
Encompass_Preferred
A pyrethroid acaricide; induces tail currents in AmNav1 and VdNav1 channels expressed Xenopus oocytes (EC50s = 60 and 160 nM, respectively) with a faster tail current decay in VdNav1 channels; topical application at 2 µl in combination with coumaphos, atrazine, 2,4-DMPF, chlorpyriphos, and chlorothalonil does not affect honeybee queen mass, egg-laying patterns, or the mass of daughter worker bees at emergence
Catalog No. 50-294-7149 Supplier Cayman Chemical Supplier No. 34520100 MG
A pyrethroid acaricide; induces tail currents in AmNav1 and VdNav1 channels expressed Xenopus oocytes (EC50s = 60 and 160 nM, respectively) with a faster tail current decay in VdNav1 channels; topical application at 2 µl in combination with coumaphos, atrazine, 2,4-DMPF, chlorpyriphos, and chlorothalonil does not affect honeybee queen mass, egg-laying patterns, or the mass of daughter worker bees at emergence