Learn More
Creative Biomart Recombinant Canine H3N2 Antigen, His-tagged
Catalog No. 502084828
Encompass_Preferred
H3N2 Canine produced in E. coli. is a single non-glycosylated polypeptide chain containing 335 amino acids (18-344) and having a molecular mass of 36.8kDa.H3N2 Canine is fused to a 6 amino acid His-ta
Catalog No. 50-208-4828 Supplier Creative Biomart Supplier No. TGO450
H3N2 Canine produced in E. coli. is a single non-glycosylated polypeptide chain containing 335 amino acids (18-344) and having a molecular mass of 36.8kDa.H3N2 Canine is fused to a 6 amino acid His-ta