Learn More
Creative Biomart Recombinant T. gondii p30 Antigen, GST-tagged
Supplier:  Creative Biomart TGO453
Encompass_Preferred
The E. coli derived recombinant protein contains the p30 (SAG1) immunodominant regions, amino acids 45-198 and fused to a 26 kDa GST tag, having a total Mw of 42.3 kDa.
Catalog No. 50-208-4831