Learn More
Creative Biomart Recombinant T. gondii p40 Antigen, His-tagged
Supplier:  Creative Biomart TGO454
Encompass_Preferred
The E. coli derived recombinant Toxoplasma Gondii p40 having a Mw of 35 kDa and fused to a 6 His tag at C-terminus can be used to test the specific IgG and IgM antibody for the diagnosis of Toxoplasma
Catalog No. 50-208-4832