Learn More
Thermco Products Inc T HANDLE WATERPROOF DIG THERMO
Catalog No. NC0992407
Encompass_Preferred
Range: -50° to 150°C (-58°/302°F).4 1/2” Stainless Steel Stem w/Probe Cover.Water ResistantNIST Traceable w/Certificate.C/F Switchable.Resolution: 0.1°.Accuracy: ± 1.0°C between -20° to 120°C.
Catalog No. NC0992407 Supplier Thermco Products Inc Supplier No. ACC610DIG
Range: -50° to 150°C (-58°/302°F).4 1/2” Stainless Steel Stem w/Probe Cover.Water ResistantNIST Traceable w/Certificate.C/F Switchable.Resolution: 0.1°.Accuracy: ± 1.0°C between -20° to 120°C.