Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ T-Link™ Clear-Vision Cassette Lens (7 Layer Set)
SDP

Catalog No. 17000118
Encompass_Preferred
Change view
Click to view available options
Product Type: Replacement Lens; For Use With (Equipment): T-Link™ Respirators
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-118 Product Type: Replacement Lens; For Use With (Equipment): T-Link™ Respirators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CA Prop 65 Free
Catalog No. 17-000-118 Supplier RPB™ Supplier No. 17817

missing translation for 'validMainframeMsg'

CA Prop 65 Free

Clear-vision cassette lens provides wearer's with a clear field of view.

  • Replacement part for T-Link™ respirators

Specifications

Color Transparent
Disposable Single-use
Material PETG
Product Type Replacement Lens
Size One Size
For Use With (Equipment) T-Link™ Respirators
Model 17-817
Dimensions (L x W x H) 10 x 2 x 14 in.
Weight (English) 1 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.