Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ T-Link™ Tychem™ 4000 SL Hood with Sealed Seams
SDP

Catalog No. 17000116
Encompass_Preferred
Change view
Click to view available options
Product Type: Hood; For Use With (Equipment): T-Link™ and T200™ Respirators
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-116 Product Type: Hood; For Use With (Equipment): T-Link™ and T200™ Respirators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-000-116 Supplier RPB™ Supplier No. 17713

missing translation for 'validMainframeMsg'

The T-Link™ Tychem™ 4000 SL Hood with Sealed Seams is a replacement part for T-Link™ and T200™ Respirators.

  • Replacement part for T-Link™ and T200™ Respirators

Specifications

Color White
Material Tychem 4000
Product Type Respirator Hood
Size One Size
For Use With (Equipment) T-Link™ and T200™ Respirators
Model 17-713
Dimensions (L x W x H) 14 x 13 x 4.5 in.
Weight (English) 0.5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.